4-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(trifluoromethyl)picolinate

A248286-100MG A248286-1G A248286-250MG
77031-442EA 29.71 USD
77031-442 77031-444 77031-446
4-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester 97%
4-(Trifluoromethyl)pyridine-2-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MW: 205.134 g/mol
MDL Number: MFCD11113409
CAS Number: 455941-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR