3-Iodo-1H-pyrazolo[3,4-c]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Iodo-6-azaindazole

A247865-100MG A247865-1G A247865-250MG A247865-5G
76987-822EA 21.22 USD
76987-822 76987-824 76987-826 76987-828
3-Iodo-1H-pyrazolo[3,4-c]pyridine 98%
3-Iodo-1H-pyrazolo[3,4-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₄IN₃
MW: 245.021 g/mol
MDL Number: MFCD11845480
CAS Number: 1082040-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR