Methyl (2E)-pent-2-enoate 95%

Supplier: Ambeed
Danger

Synonyms: Methyl trans-2-pentenoate, (E)-Methyl pent-2-enoate

A247824-1G A247824-250MG A247824-5G
76987-816EA 74.27 USD
76987-816 76987-818 76987-820
Methyl (2E)-pent-2-enoate 95%
Methyl (2E)-pent-2-enoate

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.144 g/mol
MDL Number: MFCD00137611
CAS Number: 15790-88-2
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR