3,4,5-Trifluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A247726-100G A247726-10G A247726-1G A247726-25G A247726-5G
77000-602EA 87 USD
77000-602 77000-604 77000-606 77000-608 77000-610
3,4,5-Trifluorobenzaldehyde 98%
3,4,5-Trifluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₃O
MW: 160.1 g/mol
MDL Number: MFCD00083523
CAS Number: 132123-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR