Methyl 5-chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A247719-100MG
A247719-1G
A247719-250MG
77013-920EA
20.16
USD
77013-920
77013-922
77013-924
Methyl 5-chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylate 97%
Methyl 5-chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₇ClN₂O₂ MW: 210.62 g/mol |
MDL Number:
MFCD12963479 CAS Number: 1190321-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...