4-Ethoxyphenol 98%

Supplier: Ambeed
Warning

A247672-10G A247672-25G A247672-5G A247672-100G
77031-416EA 10.61 USD
77031-416 77031-418 77031-420 77672-212
4-Ethoxyphenol 98%
4-Ethoxyphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00002334
CAS Number: 622-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR