N,N'-Bis[2-(dimethylamino)ethyl]-1,8:4,5-naphthalenetetracarboxdiimide 95+%

Supplier: Ambeed
Warning

Synonyms: 2,7-Bis(2-(dimethylamino)ethyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone

A247438-1G A247438-250MG
77013-914EA 96.55 USD
77013-914 77013-916
N,N'-Bis[2-(dimethylamino)ethyl]-1,8:4,5-naphthalenetetracarboxdiimide 95+%
N,N'-Bis[2-(dimethylamino)ethyl]-1,8:4,5-naphthalenetetracarboxdiimide

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₄O₄
MW: 408.46 g/mol
MDL Number: MFCD27923061
CAS Number: 22291-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR