5-Fluoro-3-nitrobenzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A247380-100MG A247380-250MG A247380-1G
76987-806EA 75 USD
76987-806 76987-808 77885-647
5-Fluoro-3-nitrobenzene-1,2-diamine 95%
5-Fluoro-3-nitrobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₆H₆FN₃O₂
MDL Number: MFCD26743427
CAS Number: 170098-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR