3-Methoxy-5-methylbenzoic acid 97%

Supplier: Ambeed
Warning

A247215-10G A247215-1G A247215-250MG A247215-25G
77031-394EA 215.37 USD
77031-394 77031-396 77031-398 77031-400
3-Methoxy-5-methylbenzoic acid 97%
3-Methoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C9H10O3
MW: 166.1700 g/mol
MDL Number: MFCD12405314
CAS Number: 62089-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR