N-(2-Methoxybenzyl)propan-1-amine 95+%

Supplier: Ambeed
Warning

A247116-1G A247116-5G
77013-910EA 23 USD
77013-910 77013-912
N-(2-Methoxybenzyl)propan-1-amine 95+%
N-(2-Methoxybenzyl)propan-1-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO
MDL Number: MFCD04503211
CAS Number: 807343-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR