2-tert-Butyl 6-methyl 3,4-dihydroisoquinoline-2,6(1H)-dicarboxylate 96%

Supplier: Ambeed
Warning

A247042-100MG A247042-1G A247042-250MG A247042-5G
77031-374EA 28.65 USD
77031-374 77031-376 77031-378 77031-380
2-tert-Butyl 6-methyl 3,4-dihydroisoquinoline-2,6(1H)-dicarboxylate 96%
2-tert-Butyl 6-methyl 3,4-dihydroisoquinoline-2,6(1H)-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₄
MW: 291.34 g/mol
MDL Number: MFCD08275038
CAS Number: 170097-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR