2-Chloro-4-methoxy-6-methylpyridine 95%
Supplier: Ambeed
|
Warning
|
A247034-100MG
A247034-1G
A247034-250MG
76987-796EA
63
USD
76987-796
76987-798
76987-800
2-Chloro-4-methoxy-6-methylpyridine 95%
2-Chloro-4-methoxy-6-methylpyridine
Precaución: for Research Use Only
Formula:
C₇H₈ClNO |
MDL Number:
MFCD16610591 CAS Number: 1227578-45-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...