Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine 97%

Supplier: Ambeed
Warning

Synonyms: ((2,4,6-Tri-isopropyl)phenyl)dicyclohexylphosphine

A246979-1G A246979-250MG A246979-25G A246979-5G
76987-792EA 23 USD
76987-792 76987-794 77885-644 77885-645
Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine 97%
Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine

Precaución: for Research Use Only

Formula: C₂₇H₄₅P
MDL Number: MFCD11656073
CAS Number: 303111-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR