2-Chloro-3,4-difluoroaniline 95%

Supplier: Ambeed
Warning

A246716-1G A246716-250MG A246716-5G A246716-10G
76987-780EA 35 USD
76987-780 76987-782 76987-784 77885-643
2-Chloro-3,4-difluoroaniline 95%
2-Chloro-3,4-difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄ClF₂N
MDL Number: MFCD11044823
CAS Number: 36556-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR