Methyl 5-bromo-1-methyl-1H-pyrazole-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A246677-100MG
A246677-1G
A246677-250MG
A246677-5G
77013-900EA
32.89
USD
77013-900
77013-902
77013-904
77550-582
Methyl 5-bromo-1-methyl-1H-pyrazole-3-carboxylate 97%
Methyl 5-bromo-1-methyl-1H-pyrazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₇BrN₂O₂ MW: 219.03 g/mol |
MDL Number:
MFCD22373310 CAS Number: 1222174-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...