6-Chlorobenzo[d]isoxazole-3-carboxylic acid 96%

Supplier: Ambeed
Warning

A246450-100MG A246450-250MG
77013-896EA 39.26 USD
77013-896 77013-898
6-Chlorobenzo[d]isoxazole-3-carboxylic acid 96%
6-Chlorobenzo[d]isoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₃
MW: 197.5800 g/mol
MDL Number: MFCD09027540
CAS Number: 28691-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR