5-(2-Methoxyethoxy)pyridazin-3(2H)-one 95%

Supplier: Ambeed
Warning

A246342-100MG A246342-250MG
76987-776EA 37 USD
76987-776 76987-778
5-(2-Methoxyethoxy)pyridazin-3(2H)-one 95%
5-(2-Methoxyethoxy)pyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₃
MDL Number: MFCD19704367
CAS Number: 1346697-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR