TMPyP [=alpha,beta,gamma,delta-tetrakis(1-methylpyridinium-4-yl)porphyrin-p-toluenesulfonate] 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
TMPyP4 tosylate, alpha,beta,gamma,delta-Tetrakis(1-methylpyridinium-4-yl)porphyrin p-Toluenesulphonate, 5,10,15,20-Tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulphonate)
A246194-1MG
A246194-100MG
A246194-1G
76987-774EA
6
USD
76987-774
77885-636
77885-637
TMPyP [=alpha,beta,gamma,delta-tetrakis(1-methylpyridinium-4-yl)porphyrin-p-toluenesulfonate] 98%
TMPyP [=alpha,beta,gamma,delta-tetrakis(1-methylpyridinium-4-yl)porphyrin-p-toluenesulfonate]
Precaución: for Research Use Only
Formula:
C₇₂H₇₄N₈O₁₂S₄ MW: 1371.66 g/mol |
MDL Number:
MFCD00013468 CAS Number: 36951-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...