4-Fluoro-o-anisidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-methoxyaniline hydrochloride

A246192-1G A246192-5G
76987-770EA 39.26 USD
76987-770 76987-772
4-Fluoro-o-anisidine hydrochloride 98%
4-Fluoro-o-anisidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClFNO
MW: 177.6000 g/mol
MDL Number: MFCD09800850
CAS Number: 178671-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR