3-Bromo-2,2-bis(bromomethyl)propanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-2,2-bis(bromomethyl)-1-propanol, Tribromoneopentyl alcohol, Pentaerythritol tribromide, 2,2,2-Tris(bromomethyl)ethanol
A245873-1000G
A245873-100G
A245873-500G
A245873-25G
77000-586EA
103.97
USD
77000-586
77000-588
77000-590
77304-370
3-Bromo-2,2-bis(bromomethyl)propanol 98%
3-Bromo-2,2-bis(bromomethyl)propanol
Precaución: for Research Use Only
Formula:
C5H9Br3O MW: 324.8364 g/mol |
MDL Number:
MFCD00021982 CAS Number: 1522-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...