Fmoc-Arg(Me)2-OH 95%

Supplier: Ambeed
Warning

A245359-100MG A245359-1G A245359-250MG
77000-580EA 86 USD
77000-580 77000-582 77000-584
Fmoc-Arg(Me)2-OH 95%
Fmoc-Arg(Me)2-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₈N₄O₄
MDL Number: MFCD02684510
CAS Number: 268564-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR