1-(3-Methoxyphenyl)ethanamine 98%

Supplier: Ambeed
Danger

A245336-1G A245336-5G A245336-25G
77013-874EA 18.04 USD
77013-874 77013-876 77683-736
1-(3-Methoxyphenyl)ethanamine 98%
1-(3-Methoxyphenyl)ethanamine

Precaución: For research use only.

Formula: C₉H₁₃NO
MDL Number: MFCD00673998
CAS Number: 62409-13-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR