Methyl-2,6-dimethoxypyridine-3-carboxylate 97%

Supplier: Ambeed
Warning

A245272-100MG A245272-1G A245272-250MG
76987-752EA 8 USD
76987-752 76987-754 76987-756
Methyl-2,6-dimethoxypyridine-3-carboxylate 97%
Methyl-2,6-dimethoxypyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄
MDL Number: MFCD15144586
CAS Number: 65515-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR