(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-4-phenyl-D-Phe-OH, N-Boc-3-(4-biphenylyl)-D-alanine

A245143-10G A245143-1G A245143-250MG A245143-5G A245143-25G
77000-572EA 121 USD
77000-572 77000-574 77000-576 77000-578 77881-652
(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%
(R)-3-([1,1'-Biphenyl]-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO₄
MW: 341.41 g/mol
MDL Number: MFCD00191185
CAS Number: 128779-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR