5-Bromo-2-(methylsulfinyl)pyrimidine 95+%

Supplier: Ambeed
Warning

A245047-100MG A245047-1G A245047-250MG A245047-5G
76987-740EA 12 USD
76987-740 76987-742 76987-744 76987-746
5-Bromo-2-(methylsulfinyl)pyrimidine 95+%
5-Bromo-2-(methylsulfinyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂OS
MDL Number: MFCD19689658
CAS Number: 79685-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR