O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate 97%

Supplier: Ambeed
Danger

Synonyms: N-(6-cyano-2-methyl-7-oxo-4,8-dioxa-2,5-diazadec-5-en-3-ylidene)-N-methylmethanaminium tetrafluoroborate, TOTU

A245042-10G A245042-1G A245042-25G A245042-5G
77000-564EA 20.16 USD
77000-564 77000-566 77000-568 77000-570
O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate 97%
O-[(Ethoxycarbonyl)cyanomethyleneamino]-N,N,N',N'-tetramethyluronium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₀H₁₇BF₄N₄O₃
MW: 328.07 g/mol
MDL Number: MFCD00192127
CAS Number: 136849-72-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR