PF-8380 98+%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichlorobenzyl 4-(3-oxo-3-(2-oxo-2,3-dihydrobenzo[d]oxazol-6-yl)propyl)piperazine-1-carboxylate, 3,5-dichlorobenzyl4-(3-oxo-3-(2-oxo-2,3-dihydrobenzo[d]oxazol-6-yl)propyl)piperazine-1-carboxylate

A244971-10MG A244971-1MG A244971-25MG A244971-5MG
76987-732EA 48 USD
76987-732 76987-734 76987-736 76987-738
PF-8380 98+%
PF-8380

Precaución: for Research Use Only

Formula: C₂₂H₂₁Cl₂N₃O₅
MW: 478.33 g/mol
MDL Number: MFCD20527274
CAS Number: 1144035-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR