2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%

Supplier: Ambeed
Warning

A244939-100MG A244939-1G A244939-250MG A244939-5G
77013-860EA 9 USD
77013-860 77013-862 77013-864 77013-866
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97%
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₂H₁₅BCl₂O₃
MDL Number: MFCD20526385
CAS Number: 1003298-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR