(2R)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-Boc-Piperazine-2-carboxylic acid

A244920-1G A244920-250MG A244920-25G A244920-5G
77000-556EA 62.6 USD
77000-556 77000-558 77000-560 77000-562
(2R)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 97%
(2R)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₄
MW: 230.261 g/mol
MDL Number: MFCD02179106
CAS Number: 278788-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR