5-Nitro-1H-benzo[d]imidazole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A244846-1G A244846-250MG
77031-338EA 223.85 USD
77031-338 77031-340
5-Nitro-1H-benzo[d]imidazole-2-carboxylic acid 97%
5-Nitro-1H-benzo[d]imidazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅N₃O₄
MW: 207.14 g/mol
MDL Number: MFCD09832097
CAS Number: 73903-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR