Methyl-4-amino-3-bromo-5-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-amino-3-bromo-5-nitrobenzoate

A244682-100MG A244682-1G A244682-250MG A244682-5G
77000-544EA 6.37 USD
77000-544 77000-546 77000-548 77000-550
Methyl-4-amino-3-bromo-5-nitrobenzoate 98%
Methyl 4-amino-3-bromo-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O₄
MW: 275.06 g/mol
MDL Number: MFCD08443938
CAS Number: 105655-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR