(R)-1-N-CBZ-2-Methyl-piperazine 96%

Supplier: Ambeed
Warning

Synonyms: (R)-1-Cbz-2-methylpiperazine

A244647-1G A244647-250MG A244647-25G A244647-5G
76987-720EA 32.89 USD
76987-720 76987-722 76987-724 76987-726
(R)-1-N-CBZ-2-Methyl-piperazine 96%
(R)-1-N-CBZ-2-Methyl-piperazine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.294 g/mol
MDL Number: MFCD04115051
CAS Number: 923565-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR