Fmoc-L-homophenylalanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-HoPhe-OH, (S)-2-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-phenylbutyric acid, Fmoc-Homophe-OH, N-Fmoc-L-homophenylalanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-homophenylalanine, (S)-2-(Fmoc-amino)-4-phenylbutyric acid
A244629-100G
A244629-1G
A244629-25G
A244629-5G
A244629-10G
77000-540EA
114
USD
77000-540
77000-542
77550-572
77568-380
77881-649
Fmoc-L-homophenylalanine 97%
Fmoc-L-homophenylalanine
Precaución: For research use only.
Formula:
C₂₅H₂₃NO₄ |
MDL Number:
MFCD00077046 CAS Number: 132684-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...