Potassium-2-ethylhexanoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Potassium 2-ethylhexanoate, 2-Ethylhexanoic Acid Potassium Salt
A244624-1000G
A244624-100G
A244624-25G
A244624-500G
A244624-5G
77000-530EA
93
USD
77000-530
77000-532
77000-534
77000-536
77000-538
Potassium-2-ethylhexanoate 98%
Potassium-2-ethylhexanoate
Precaución: for Research Use Only
Formula:
C₈H₁₆O₂ MW: 182.30 g/mol |
MDL Number:
MFCD00045896 CAS Number: 3164-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...