3-Iodo-5-methyl-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Warning

A244576-1G A244576-250MG
77000-524EA 143.23 USD
77000-524 77000-526
3-Iodo-5-methyl-1H-pyrrolo[2,3-b]pyridine 98%
3-Iodo-5-methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₇IN₂
MDL Number: MFCD11857743
CAS Number: 1138443-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR