4'-Methoxy-2'-(trifluoromethyl)acetophenone 97%

Supplier: Ambeed
Warning

A244482-25G A244482-5G A244482-250MG A244482-1G
76987-710EA 201 USD
76987-710 76987-712 77612-362 77890-398
4'-Methoxy-2'-(trifluoromethyl)acetophenone 97%
4'-Methoxy-2'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MDL Number: MFCD01091009
CAS Number: 220141-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR