Lauric acid 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Dodecanoic acid, Dodecylic acid
A244321-25G
A244321-100G
A244321-500G
A244321-1000G
77000-522EA
5
USD
77000-522
77568-376
77600-978
77881-648
Lauric acid 98%
Lauric acid
Precaución: For research use only.
Formula:
C₁₂H₂₄O₂ MW: 200.32 g/mol Boiling Pt: 296 |
MDL Number:
MFCD00002736 CAS Number: 143-07-7 UN: 3077 ADR: 9 Ⅲ Merck Index: 13,05400 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...