1-Propyl-1H-pyrazole-4-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
A244131-100MG
A244131-250MG
77000-518EA
20.16
USD
77000-518
77000-520
1-Propyl-1H-pyrazole-4-carbaldehyde 97%
1-Propyl-1H-pyrazole-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂O MW: 138.17 g/mol |
MDL Number:
MFCD03727244 CAS Number: 473249-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...