Sodium 4-(2-(2-(2-chloro-3-(2-(1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)ethylidene)cyclohex-1-en-1-yl)vinyl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate 97%
Supplier: Ambeed
|
Warning
|
A244100-100MG
A244100-1G
A244100-250MG
A244100-5G
77031-332EA
45
USD
77031-332
77031-334
77031-336
77600-970
Sodium 4-(2-(2-(2-chloro-3-(2-(1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)ethylidene)cyclohex-1-en-1-yl)vinyl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate 97%
Sodium 4-(2-(2-(2-chloro-3-(2-(1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)ethylidene)cyclohex-1-en-1-yl)vinyl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate
Precaución: for Research Use Only
Formula:
C₄₆H₅₀ClN₂NaO₆S₂ |
MDL Number:
MFCD04038859 CAS Number: 172616-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...