N-(2-Methylbenzyl)propan-2-amine 95%

Supplier: Ambeed
Warning

A243996-10G A243996-1G A243996-5G
77013-832EA 118 USD
77013-832 77013-834 77013-836
N-(2-Methylbenzyl)propan-2-amine 95%
N-(2-Methylbenzyl)propan-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₇N
MDL Number: MFCD08060639
CAS Number: 91338-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR