8-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A243963-100MG A243963-1G A243963-250MG A243963-5G
77013-822EA 7 USD
77013-822 77013-824 77013-826 77013-828
8-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
8-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁Cl₂N
MDL Number: MFCD08437644
CAS Number: 61563-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR