Ethyl-5-amino-1-(4-methylphenyl)-1H-pyrazole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-amino-1-(p-tolyl)-1H-pyrazole-4-carboxylate

A243599-10G A243599-1G A243599-5G
77000-496EA 136.86 USD
77000-496 77000-498 77000-500
Ethyl-5-amino-1-(4-methylphenyl)-1H-pyrazole-4-carboxylate 95%
Ethyl 5-amino-1-(4-methylphenyl)-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₅N₃O₂
MW: 245.2800 g/mol
MDL Number: MFCD00973936
CAS Number: 15001-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR