Pralatrexate 98%

Supplier: Ambeed
Warning

Synonyms: (2S)-2-(4-(1-(2,4-Diaminopteridin-6-yl)pent-4-yn-2-yl)benzamido)pentanedioic acid, (2S)-2-[4-[1-(2,4-Diaminopteridin-6-yl)pent-4-yn-2-yl]benzamido]pentanedioic acid

A243480-100MG A243480-250MG A243480-50MG
76987-672EA 370.26 USD
76987-672 76987-674 76987-676
Pralatrexate 98%
Pralatrexate

Precaución: for Research Use Only

Formula: C₂₃H₂₃N₇O₅
MW: 477.48 g/mol
MDL Number: MFCD00920897
CAS Number: 146464-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR