Methyl 4-(piperidin-3-yl)benzoate 95%

Supplier: Ambeed
Warning

A243304-100MG A243304-1G A243304-250MG A243304-5G
76987-662EA 190.97 USD
76987-662 76987-664 76987-666 76987-668
Methyl 4-(piperidin-3-yl)benzoate 95%
Methyl 4-(piperidin-3-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₂
MW: 219.27 g/mol
MDL Number: MFCD09055085
CAS Number: 927801-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR