L(+)-Norleucine 97%

Supplier: Ambeed
Warning

Synonyms: H-Nle-OH, H-NorLeu-OH, (S)-2-Aminohexanoic acid, (S)-(+)-Norleucine

A243254-100G A243254-10G A243254-1G A243254-25G A243254-5G
77000-488EA 151.71 USD
77000-488 77000-490 77000-492 77600-958 77600-960
L(+)-Norleucine 97%
L(+)-Norleucine

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
MDL Number: MFCD00064423
CAS Number: 327-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR