H-Cys(pMeOBzl)-OH 95%

Supplier: Ambeed
Warning

Synonyms: S-(4-Methoxybenzyl)-L-cysteine, H-Cys(Mob)-OH

A243028-100G A243028-1G A243028-25G A243028-5G
77000-482EA 269 USD
77000-482 77000-484 77000-486 77672-174
H-Cys(pMeOBzl)-OH 95%
H-Cys(pMeOBzl)-OH

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃S
MW: 241.305 g/mol
MDL Number: MFCD00037207
CAS Number: 2544-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR