2-(Isochroman-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

A242642-100MG A242642-1G A242642-250MG
77031-302EA 21 USD
77031-302 77031-304 77031-306
2-(Isochroman-1-yl)acetic acid 95%
2-(Isochroman-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MDL Number: MFCD05721361
CAS Number: 22901-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR