N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: Z-Tyr(tBu)-OH

A242603-100G A242603-10G A242603-25G A242603-5G
76987-654EA 160 USD
76987-654 76987-656 76987-658 76987-660
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine 98%
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine

Precaución: for Research Use Only

Formula: C₂₁H₂₅NO₅
MDL Number: MFCD00065701
CAS Number: 5545-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR