6-Bromo-2-methyl-1,2,3,4-tetrahydroquinoline 95%

Supplier: Ambeed
Warning

A242583-1G A242583-250MG
77013-794EA 433.91 USD
77013-794 77013-796
6-Bromo-2-methyl-1,2,3,4-tetrahydroquinoline 95%
6-Bromo-2-methyl-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrN
MW: 226.11 g/mol
MDL Number: MFCD22035739
CAS Number: 42835-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR