4'-Methyl-4-biphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4'-Methyl-[1,1'-biphenyl]-4-yl)boronic acid

A242390-1G A242390-250MG A242390-25G A242390-5G
77013-786EA 31.83 USD
77013-786 77013-788 77013-790 77013-792
4'-Methyl-4-biphenylboronic acid 97%
4'-Methyl-4-biphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BO₂
MW: 212.06 g/mol
MDL Number: MFCD04038170
CAS Number: 393870-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR